To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Host:
- Primary antibody reactivity:Human
- Description:Human Perforin ELISA Assay Kit
- Size:1 kit
- Environmentally Preferable:
- Cat. No.:102994-848
- Supplier no.:PFN31-K01
Human Perforin ELISA to determine Perforin in supernatant, buffered solutions.
- Dynamic Range: 62.5-2000 pg/ml
- Sensitivity: 40 pg/ml
- Incubation: 2h45
The Eagle Biosciences Human Perforin ELISA Assay Kit is to be used for the in-vitro quantitative determination of Perforin in supernatant, buffered solutions. The assay will recognize natural Perforin.