Order Entry
Puerto Rico
ContactUsLinkComponent
Kallikrein-related peptidase 8 (KLK8) ELISA, Eagle Biosciences, Inc.
Kallikrein-related peptidase 8 (KLK8) ELISA, Eagle Biosciences, Inc.
Catalog # 102993-728
Supplier:  Eagle Biosciences
CAS Number:  
Kallikrein-related peptidase 8 (KLK8) ELISA, Eagle Biosciences, Inc.
Catalog # 102993-728
Supplier:  Eagle Biosciences
Supplier Number:  KL831-K01
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Kallikrein-related peptidase 8 (KLK8) ELISA
  • Size:
    1 kit
  • Cat. No.:
    102993-728
  • Supplier no.:
    KL831-K01

Specifications

About this item

Kallikrein-Related Peptidase 8 (KLK8) ELISA detects the concentration of KLK8 in human serum.

  • Dynamic Range: 1 - 256 ng/ml
  • Sensitivity: 1 ng/ml
  • Incubation: 2.5 hours

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK8 is one of the fifteen kallikrein subfamily member. The KLK8 enzyme exhibits distinct patterns of expression that suggest roles in brain plasticity and ovarian cancer.