Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Primary antibody reactivity:Human
- Target protein:IL-4
- Description:Human CD124 / IL-4R ELISA Assay Kit
- Size:1 kit
- Cat. No.:102994-036
- Supplier no.:C2431-K01
Human IL-4R ELISA detects soluble IL-4 receptor molecule (CD124) in various samples.
- Dynamic Range: 31.25-1000 pg/ml
- Sensitivity: 11.5 pg/ml
- Incubation: 1h45
The Eagle Biosciences Human IL-4R ELISA is to be used for the in-vitro quantitative determination of soluble IL-4 receptor molecule (CD124) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human IL-4R. The Eagle Biosciences Human IL-4R ELISA Assay kit is for Research Use Only.