Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugate ELISA:HRP-Streptavidin
- Primary antibody reactivity:Human
- Target protein:Granzyme B
- Description:Human Granzyme B ELISA Assay Kit
- Size:1 kit
- Cat. No.:102994-426
- Supplier no.:GRB31-K01
Specifications
About this item
CD44 ELISA detects soluble CD44 in human serum, plasma, buffered solutions or cell culture medium.
- Dynamic Range: 31.25-1000 pg/ml
- Sensitivity: 20 pg/ml
- Incubation: 3h00
The Eagle Biosciences Human Granzyme B ELISA is to be used for the in-vitro quantitative determination of Granzyme B in human serum, plasma, buffered solutions or cell culture medium. The Eagle Biosciences Human Granzyme B ELISA Assay kit is for Research Use Only.