Order Entry
United States
ContactUsLinkComponent
 
Osteocalcin ELISA, Eagle Biosciences
  102994-826
 :  
Osteocalcin ELISA, Eagle Biosciences
  102994-826
 :  OST31-K01
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Host:
  • Primary antibody reactivity:
  • Description:
    Osteocalcin ELISA
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    102994-826
  • Supplier no.:
    OST31-K01

 

 

Osteocalcin ELISA detects human osteocalcin (1-49) and osteocalcin (1-43) levels.

  • Dynamic Range: 4.0 - 64 ng/ml
  • Sensitivity: .31 ng/ml
  • Incubation: 1.5 hours

The Eagle Biosciences Human Osteocalcin ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of both human osteocalcin (1-49) and osteocalcin (1-43) (also referred as N-terminal & mid-regional osteocalcin) levels in test samples). The Eagle Biosciences Human Osteocalcin ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.