Order Entry
Puerto Rico
ContactUsLinkComponent
Human IL-6 HS ELISA Assay Kit, Eagle Biosciences, Inc.
Human IL-6 HS ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 102993-514
Supplier:  Eagle Biosciences
CAS Number:  
Human IL-6 HS ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 102993-514
Supplier:  Eagle Biosciences
Supplier Number:  IL639-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-6
  • Description:
    Human IL-6 HS ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102993-514
  • Supplier no.:
    IL639-K01

Specifications

About this item

Human High Sensitivity IL-6 ELISA detects IL-6 in supernatants,buffered solutions,serum,plasma.

  • Dynamic Range: 1.56-50pg/ml
  • Sensitivity: 0.8 pg/ml
  • Incubation: 3h45

Interleukin-6 (IL-6) is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis and may play a central role in host defense mechanisms (13, 31). The gene for human IL-6 has been localized to chromosome 7p21 (1). The genomic sequence has been determined (36). IL-6 is usually not produced constitutively by normal cells, but its expression is readily induced by a variety of cytokines (28), lipopolysaccharide (25) or viral infections (3). The IL-6 gene product is a single chain protein with a molecular mass ranging from 21 to 28 kDa, depending on the cellular source. Extensive posttranslational modifications like N- and O-linked glycosylation (20) as well as phosphorylation (21) seem to account for this heterogeneity. The cDNA for IL-6 predicts a precursor protein of 212 amino acids (10). IL-6 is a pleiotropic cytokine produced by a variety of cells. It acts on a wide range of tissues, exerting growth-induction, growth-inhibition, and differentiation respectively, depending on the nature of the target cells. The abnormal production of IL-6 was first suggested to be related to polyclonal B-cell activation with autoantibody production in patients with cardiac myxoma (9). Since then, IL-6 has been suggested to be involved in the pathogenesis of a variety of diseases. Measurement of IL-6 levels in serum and other body fluids thus provides more detailed insights into various pathological situations.