Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugate ELISA:HRP
- Description:Fetuin A ELISA
- Size:1 kit
- Cat. No.:102994-386
- Supplier no.:FET39-K01
Fetuin A ELISA detects human Fetuin-A, in serum, plasma, cell culture supernatant, tissue and urine.
- Dynamic Range: 12.5 - 370 ng/ml
- Sensitivity: 5.0 ng/mL
- Incubation: 3 hours
Fetuin-A, also known as alpha-2-HS glycoprotein, is a 59 kDa glycoprotein that consists of two amino-terminal cystatin domains and a smaller carboxyl-terminal domain. Fetuin-A is synthesized by the liver and secreted into blood stream, where its concentration in adult mammals ranges from 0.5 – 1.0 g/L. Fetuin-A occurs in high serum concentration during fetal life and involves in protease inhibitory activities and development-associated regulation of calcium metabolism and osteogenesis. It accumulates in bones and teeth as a major fraction of noncollagenous bone proteins.