Order Entry
United States
ContactUsLinkComponent
IGFBP-2 ELISA, Eagle Biosciences
IGFBP-2 ELISA, Eagle Biosciences
Catalog # 102994-276
Supplier:  Eagle Biosciences
CAS Number:  
IGFBP-2 ELISA, Eagle Biosciences
Catalog # 102994-276
Supplier:  Eagle Biosciences
Supplier Number:  E05
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
  • Description:
    IGFBP-2 ELISA
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    102994-276
  • Supplier no.:
    E05

Specifications

About this item

IGFBP-2 ELISA detects human IGFBP-2 in biological samples

  • Dynamic Range: 2 -80 ng/ml
  • Sensitivity: 0.2 ng/ml
  • Incubation: 2 hours

Insulin-like growth factors (IGFs) regulate the proliferation, differentation, apoptose, cell adhesion and metabolism in various tissues and cell types. The IGF-I, which is produced mainly in liver under the influence of growth hormone (GH), regulates as hormone the linear growth of the bones and the process of sexual maturity, while IGF-II is mainly a growth factor of fetal tissue. The biological actions of IGF over the IGF-Type-I receptor are modulated variably through the IGF binding proteins (IGFBP-1 to-6). IGFBP-2 is, after IGFBP-3, the second most frequent IGFBP in the human blood. IGFs, especially tumor typical pro-IGF-forms and hormones regulate the expression of IGFBP-2, GH effect is thereby inhibiting. At cellular level IGFBP-2 seems to stimulate the proliferation and dissemination of solid tumors via an IGF-independent mechanism.

For Research Use Only.