Order Entry
United States
ContactUsLinkComponent
hGH Sensitive ELISA , Eagle Biosciences, Inc.
hGH Sensitive ELISA , Eagle Biosciences, Inc.
Catalog # 102994-264
Supplier:  Eagle Biosciences
CAS Number:  
hGH Sensitive ELISA , Eagle Biosciences, Inc.
Catalog # 102994-264
Supplier:  Eagle Biosciences
Supplier Number:  E022
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
  • Description:
    hGH Sensitive ELISA
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    102994-264
  • Supplier no.:
    E022

Specifications

About this item

hGH Sensitive ELISA is quantitatively measures levels of human growth hormone in serum and plasma.

  • Dynamic Range: 0.05 - 1.0 ng/mL
  • Sensitivity: 12 pg/ml
  • Incubation: 2.5 hours

The Endocrinology system of human Growth Hormone (hGH), also named Somatropin, is characterized by an extreme complexity. hGH is the product of the GH-1 gene located on chromosome 17 and expressed in pituitary cells. 80% of the gene expression results in a non-glycolsylated 22 kDa protein consisting of 191 amino acids. The other 20% of gene expression results in a variant form of 20 kDa by alternative splicing. Additionally, several smaller variants can be found in circulation as well as translational modified proteins and different degrees of protein aggregation. Further on, bioactivity of Growth Hormone is regulated by a specific binding protein (GHBP) formed by the extra cellular part of the cellular transmembran GH-receptor. These modifications allow a tight control of the half-life period hGH and of its bioactivity.