Order Entry
Puerto Rico
ContactUsLinkComponent
TNF alpha Neutralization Rate/Infliximab ELISA Assay Kit, Eagle Biosciences
TNF alpha Neutralization Rate/Infliximab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-028
Supplier:  Eagle Biosciences
CAS Number:  
TNF alpha Neutralization Rate/Infliximab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-028
Supplier:  Eagle Biosciences
Supplier Number:  R444
CAS Number:  

Specifications

  • Target protein:
    TNF-alpha
  • Description:
    TNF alpha Neutralization Rate/Infliximab ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    103389-028

Specifications

About this item

recoveryELISA RTI is for research use only and not to be used in diagnostic procedures.

  • Dynamic Range: Infliximab: 0.1 – 15 µg/mL; TNF alpha in sample without Infliximab: 2-400ng/ml;TNF alpha in sample with Infliximab: 1-160ng/ml
  • Sensitivity: Infliximab: 0.1 µg/mL; TNF alpha in sample without Infliximab: 2ng/ml;TNF alpha in sample with Infliximab: 1ng/ml
  • Incubation: overnight

Tumour necrosis factor (TNFα) is a signalling substance of the immune system which can cause, among other things, the formation of inflammatory mediators in cells. In inflammatory diseases such as rheumatoid arthritis, there is overproduction of TNFα, which can be inhibited by using therapeutic antibodies.Infliximab is an chimeric monoclonal antibody which is used as a TNFα inhibitor. Infliximab binds specifically to the TNFα protein and neutralises the biological effect of TNFα.Infliximab is approved for the treatment of the following diseases: rheumatoid arthritis, psoriatic arthritis, psoriasis, ankylosing spondylitis, Crohn's disease and ulcerative colitis. In therapy using Infliximab, contraindications such as hypersensitivity to Infliximab and also serious infections, sepsis, tuberculosis and cases of opportunistic infections have been noted. Weakening of the body's defence is produced by the immunosuppressive effect of Infliximab. Excessive neutralization of TNFα during treatment with Infliximab increases the risk of developing opportunistic infections. In view of this, monitoring the treatment antibody levels and degree of neutralization of TNFα would appear useful during therapy using the TNFα inhibitor Infliximab. In comparison with the tests available on the market, the recoveryELISA TNFα Neutralization Rate/Infliximab (RTI) Kit can be used to simultaneously determine the free TNFα target Antigens, the available therapeutic antibody Infliximab and its ability to neutralise TNFα.