Anti Cardiolipin Screen ELISA, Eagle Biosciences
103389-128
:
- Description:ANTI CARDIOLIPIN SCREEN ELISA
- Size:1 kit
- Cat. No.:103389-128
Anti Cardiolipin Screen ELISA Assay Kit is for research use only and not for diagnostic procedures.
- Dynamic Range: 5 - 80 AU/ml
- Sensitivity: 0.08 AU/ml
- Incubation: 2.5 hours
Eagle Biosciences Anti Cardiolipin Screen ELISA Assay Kit allows to determine the unknown concentration of auto-antibodies directed against the Cardiolipin-β2-glycoprotein complex through two different calibration curve (one specific for IgG test, one specific for IgM test), two different conjugates linked to horseradish peroxidase (one specific for IgG test, the other specific for IgM test) and one microplate only. The principle of the method and the procedure are the same in both the tests. Use reagents for IgG or reagents for IgM depending on the isotype which is under investigation.