Anti-Baculovirus gp64-(R-PE) Conjugated Mouse Monoclonal Antibody [clone: AcV1]
77705-804
New Product
- Storage Buffer:PBS
- Storage Temperature:Store at 2...8 °C, protect from light
- Cat. No.:77705-804
PE-labeled antibody directed to gp64, the baculovirus envelope protein required for cell-to-cell spread.
The baculovirus gp64 envelope glycoprotein is a major component of the envelope of the budded baculovirus and is essential for entry into cultured insect host cells. The anti-gp64 antibody is effective for direct immunofluorescence staining of insect cells for flow cytometric analysis.Applications include baculovirus titration, infection kinetics monitoring and protein expression screening.
: Standard overnight.
Type: Primary
Antigen: gp64
Clonality: Monoclonal
Clone: AcV1
Conjugation: R-PE (R-Phycoerythrin)
Host: Mouse
Isotype: IgG2a
Reactivity: Baculovirus
Storage Buffer: Phosphate buffered saline (PBS) containing 0.05% BSA and 0.09% sodium azide