Oakton T100 Replacement Turbidity Calibration Kit, Cole-Parmer
Catalog # 101076-390
Supplier: OAKTON INSTRUMENTS MS
Specifications
- Description:Oakton T100 Replacement Turbidity Calibration Kit
- Cat. No.:101076-390
- For use with:Oakton T100 Turbidity Meter
- Supplier no.:WD-35635-52
Specifications
About this item
For calibration of Oakton T-100 Turbidimeter.
Ordering information: Includes one each of 60 mL HPDE bottle of four NTU standards: 0.02, 20.0, 100 and 800 NTU.