ContactUsLinkComponent
Oakton pH 5+ and 6+ Handheld pH Meters, Cole-Parmer
Oakton pH 5+ and 6+ Handheld pH Meters, Cole-Parmer
Certificates
undefined
Oakton pH 5+ and 6+ Handheld pH Meters, Cole-Parmer
Specifications
  • Catalog No:
  • 101076-426
  • 101076-430
  • Model:
  • pH 5+
  • pH 6+
  • pH Range:
  • 0 - 14
  • 0 - 14
  • pH Resolution:
  • 0.01 or 0.1
  • 0.01 or 0.1
  • pH Accuracy:
  • ±0.01 pH
  • ±0.01 pH
  • pH Calibration:
  • Up to 5 points
  • Up to 5 points
  • mV range:
  • −1999...+1999
  • mV resolution:
  • 1
  • mV accuracy:
  • ±0.2 mV or 0.05%, whichever is greater
  • Temperature Range:
  • −10...+110 °C
  • −10...+110 °C
  • Temperature Compensation:
  • Automatic
  • Automatic
  • Display:
  • LCD
  • LCD
  • Power supply:
  • 4 AAA batteries (included)
  • 4 AAA batteries (included)
  • Dimensions:
  • 3 3/8 × 1 11/16"
  • 3 3/8 × 1 11/16"
Specifications