Order Entry
Puerto Rico
ContactUsLinkComponent
HardyDisks™ Trimethoprim, TMP-5, AST
HardyDisks™ Trimethoprim, TMP-5, AST
Catalog # 76169-772
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
HardyDisks™ Trimethoprim, TMP-5, AST
Catalog # 76169-772
Supplier:  Hardy Diagnostics
Supplier Number:  Z9181
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    Trimethoprim, TMP-5
  • Size:
    1×50 Disks
  • Cat. No.:
    76169-772
  • Application:
    Clinical

Specifications

About this item

HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.

  • Impregnated disks
  • Spring loaded dispenser
  • Smooth and reliable dispensing

Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.