CHO-ES (suspension CHO K1) cells, frozen in ESF SFM, 20M cells/vial
Catalog # 77705-766
Supplier: EXPRESSION SYSTEMS LLC
New Product
CAS Number:
Specifications
- Cat. No.:77705-766
Specifications
About this item
Expression Systems offers 293 and CHO mammalian cell lines adapted to serum-free suspension culture in ESF SFM.
- Capable of high levels of recombinant protein expression
- Good growth in adherent or suspension culture
- Quality and performance testing
- Organism: Cricetulus griseus, hamster, Chinese
- Tissue: Ovary
- Cell Type: Epithelial
Biosafety level 1: Biosafety classification is based on U.S. Public Health Service Guidelines, it is the responsibility of the customer to ensure that their facilities comply with biosafety regulations for their own country.
CHO-ES cells are derived from the CHO (Chinese Hamster Ovary) cell line. CHO-ES cells are adapted for suspension culture in ESF SFM and are available as a frozen vial or suspension culture.
The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.
Delivery: Priority overnight.