Specifications
- Description:DNA-OFF™ Cleaning Solution
- Size:500 mL
- Environmentally Preferable:
- Cat. No.:76020-300
Specifications
About this item
DNA-OFF™ is a non-alkaline cleaning solution that is highly active against DNA contamination.
- Ready to use solution
- Non-carcinogenic
- Non-corrosive
- Heat resistant
DNA-OFF™ solution contains a surfactant plus an agent that destroys DNA intended for use at PCR workstations including countertops, apparatus and pipettors
DNA-OFF™ is used for eliminating DNA from all surfaces. It contains a surfactant plus an agent that destroys DNA intended for use at PCR workstations including countertops, apparatus and pipettors. It is stable and heat resistant. In addition, the solution is non-corosive and does not contain carcignogenic compounds.
Presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts. It is stable and heat resistant. In addition, the solution is non-corrosive and does not contain carcinogenic compounds.