Cap Strips for Well Plates, Amplitube™, Simport
Catalog # 76198-306
Supplier: Simport Scientific
Specifications
- Description:Domed 8-Strip Cap, Natural
- Cat. No.:76198-306
Specifications
About this item
Amplitube™ domed cap strips ensure a perfect closure and a positive seal during thermal cycling and prevent evaporation.
- 8 domed cap strips
- Made of FDA compliant polypropylene
- Free of RNase, DNase, Pyrogen, and DNA
- Leakproof/evaporation tested
Packaging: Packaged in tamperproof resealable bags.