ContactUsLinkComponent
KIMTECH PURE® A8 Certified Lab Coat + Extra Protection, Kimberly Clark Professional
  76199-146
   Waste Reducing
KIMTECH PURE® A8 Certified Lab Coat + Extra Protection, Kimberly Clark Professional
  76199-146
 :  10049
   Waste Reducing
  • Clothing Size:
    XXL
  • Garment application:
    Laboratory
  • Gender:
  • Color:
    Blue
  • Environmentally Preferable:
    Waste Reducing
  • Cat. No.:
    76199-146
Disposable, fluid-resistant lab coats are manufactured from a protective three-layer spunbond-meltblown-spunbond (SMS) fabric that offers exceptional resistance to blood strikethrough for moderate fluid contact.
  • Breathable, three-layer SMS fabric
  • Extra Protection offers blood strikethrough resistance
  • Soft knit cuffs for added comfort and protection
  • Nonsterile
  • Latex-free
Lab coats feature a six-snap front closure, three reinforced pockets, back vent, traditional collar, and knit cuffs. Tailored styling and fit enhances worker comfort.
Recyclable through the Rightcycle Program.