ContactUsLinkComponent
Touch Test™ Sensory Probes, Stoelting
Touch Test™ Sensory Probes, Stoelting
Catalog # 95060-230
Supplier:  Stoelting
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Touch Test™ Sensory Probes, Stoelting
Catalog # 95060-230
Supplier:  Stoelting
Supplier Number:  58011
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Touch Test™ Sensory Evaluator, Kit of 20
  • Environmentally Preferable:
  • Cat. No.:
    95060-230
  • Supplier no.:
    58011
Specifications
About this item
The Semmes-Weinstein monofilaments provide an approximate logarithmic scale of actual force, and a linear scale of perceived intensity. Ideal for use in clinical settings, and can be used to diagnose pathologies of hyper or hypo-aesthesia.
When the tip of a fiber of given length and diameter is pressed against the skin at right angles, the force of application increases as long as the researcher continues to advance the probe, until the fiber bends
Rodents exhibit a paw withdrawal reflex when the paw is unexpectedly touched. Touch Test™ sensory evaluator can be used on the Plantar surfaces of the foot of a rat or mouse.
Subsets within the set of 20 probes distinguish pathologies on different parts of the body (foot, hand, lip, cheek).
After the fiber bends, continued advance creates more bend, but not more force of application. This makes it possible for the researcher using a hand-held probe to apply a reproducible force, within a wide tolerance, to the skin surface.