Surgical Face Mask, Veltek Associates
Supplier: Veltek Associates
Certificates
About this item
PF-2 face masks provide excellent barrier capabilities, eliminate fogging of goggles, and are available in multiple varieties.
- Material is soft, comfortable, and has excellent breathability
- Made of rayon and laminated with an acrylic binder that has low particulate and shedding characteristics
The PF-2 material blend is designed for use in pharmaceutical, biotechnology, and medical device industries.