SUMO-1-AMC
Catalog # 76045-558
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:50 µg
- Protein/Peptide Name:SUMO-1-AMC
- Purity:>95% (HPLC)
- Molecular Weight:~11.3kDa
- Formulation:Liquid. In 50mM HEPES, pH 7.5, containing 100 mM NaCl.
- Cat. No.:76045-558
Specifications
About this item
Produced in E. coli.
Fluorogenic substrate for deSUMOylating enzymes. SUMO1-AMC is a sensitive fluorogenic substrate for deSUMOylating enzymes, including SENP1 (Prod. No. BML-UW9760) and SENP2 (Prod. No. BML-UW9765). It is a particularly useful reagent for the study of deSUMOylating activity where detection sensitivity or continuous monitoring of activity is essential.