Human Recombinant Growth and differentiation factor-15
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli. Non-glycosylated, disulfide linked homodimer, containing two identical 113 amino acid chains.
- Carrier-free
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Endotoxin Content:≤1 EU/μg protein measured by kinetic LAL analysis.
- Biological Activity:The activity is determined by the inhibition of DU-145 cells which is typically observed at a concentration of 1-2 µg/ml.
- Protein/Peptide Name:Growth and differentiation factor-15
- Purity:>95% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Molecular Weight:~24.5 kDa
- Formulation:Lyophilized. Sterile filtered.