Human Recombinant DLL1 (Soluble)
Catalog # 76045-506
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:25 µg
- Endotoxin Content:< 0.1 ng per μg of sDLL-1.
- Biological Activity:Determined by the dose dependent growth suppression of the human acute monocytic leukemia cell line, THP-1. sDLL-1 inhibits the proliferation in THP-1 cells using a concentration of 3-5 μg/ml.
- Protein/Peptide Name:DLL1 (soluble)
- Purity:>95% (SDS-PAGE and HPLC)
- Molecular Weight:~57-60kDa
- Formulation:Lyophilized from 1X PBS, pH 7.5.
- Cat. No.:76045-506
Specifications
About this item
Notch ligand
Produced in HEK 293 cells. Recombinant human sDLL-1 contains 522 amino-acid residues.