Anti-Zinc finger protein 682 Rabbit Polyclonal Antibody
Catalog # 103286-696
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Zinc finger protein 682
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:91120
- Antigen Synonyms:zinc finger protein 682|FLJ90362|BC39498_3
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against a recombinant protein corresponding to amino acids: SCGLEDLHLRKDGENVGECKDQKEI
- Purification:Immunogen affinity purified
- Cat. No.:103286-696
- Supplier no.:NBP2-33773
Specifications
About this item
The Zinc finger protein 682 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Zinc finger protein 682. This antibody reacts with human. The Zinc finger protein 682 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Zinc finger protein 682
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human