Anti-SARS Nucleocapsid Protein Rabbit Polyclonal Antibody
103358-384
- Antibody Type:Primary
- Antigen Name:SARS Nucleocapsid Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 ml
- Antigen Synonyms:SARS coronavirus nucleocapsid protein|N protein|SARS coronavirus N protein|N structural protein|SARSCoV N protein|SARS N protein|SARSCoV nucleocapsid protein|N|NC|SARS CoV|SARS CoV nucleocapsid protein|Nucleocapsid protein|SARS CoV N protein|Severe acute respiratory syndrome|SARSCoV
- Storage Buffer:Whole antisera with PBS and 0.2% Gelatin
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 20-34 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
- Purification:Unpurified
- Cat. No.:103358-384
The SARS Nucleocapsid Protein Antibody from Novus Biologicals is a rabbit polyclonal antibody to SARS Nucleocapsid Protein. This antibody reacts with human. The SARS Nucleocapsid Protein Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: SARS Nucleocapsid Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human