Anti-SARS Nucleocapsid Protein Rabbit Polyclonal Antibody
103360-440
- Antibody Type:Primary
- Antigen Name:SARS Nucleocapsid Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus,Human
- Western Blot:Yes
- Size:0.025 mg
- Antigen Synonyms:SARS coronavirus nucleocapsid protein|N protein|SARS coronavirus N protein|N structural protein|SARSCoV N protein|SARS N protein|SARSCoV nucleocapsid protein|N|NC|SARS CoV|SARS CoV nucleocapsid protein|Nucleocapsid protein|SARS CoV N protein|Severe acute respiratory syndrome|SARSCoV
- Storage Buffer:PBS containing 0.05% BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 399-411 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
- Purification:Protein G purified
- Cat. No.:103360-440
The SARS Nucleocapsid Protein Antibody from Novus Biologicals is a rabbit polyclonal antibody to SARS Nucleocapsid Protein. This antibody reacts with human, viral. The SARS Nucleocapsid Protein Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: SARS Nucleocapsid Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Viral