Anti-Sars Envelope Protein Rabbit Polyclonal Antibody
Catalog # 103360-332
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Sars Envelope Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Storage Buffer:PBS containing 0.2% gelatin
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 59-70 YSRVKNLNSSEG of the putative SARS envelope protein.
- Purification:Protein G purified
- Cat. No.:103360-332
Specifications
About this item
The Sars Envelope Protein Antibody from Novus Biologicals is a rabbit polyclonal antibody to Sars Envelope Protein. This antibody reacts with human. The Sars Envelope Protein Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Sars Envelope Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human