To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:RNase L
- Clonality:Monoclonal
- Clone:2E9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Primates,Hamster,Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:6041
- Antigen Synonyms:Ribonuclease 4|MGC104972|RNase L|2-5A-dependent ribonuclease|HPC1|PRCA1|ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent)|2-5A-dependent RNase|interferon-induced 2-5A-dependent RNase|Ribonuclease L|2'-5'-oligoisoadenylate synthetase-dependent|MGC133329|EC 3.1.26.-|prostate cancer 1|RNS4DKFZp781D08126
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Purified full-length recombinant human RNaseL produced in insect cells. [UniProt# Q05823]
- Purification:Protein G purified
- Cat. No.:103357-830
Specifications
About this item
The RNase L Antibody (2E9) [FITC] from Novus Biologicals is a mouse monoclonal antibody to RNase L. This antibody reacts with human, hamster, primate, mouse (negative), rat (negative). The RNase L Antibody (2E9) [FITC] has been validated for the following applications: ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: RNase L
Clonality: Monoclonal
Clone: 2E9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Hamster, Primate, Mouse (Negative), Rat (Negative)