Anti-Proteasome 19S 10B Mouse Monoclonal Antibody [clone: 1B4]
Catalog # 103308-512
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome 19S 10B
- Clonality:Monoclonal
- Clone:1B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:5708
- Antigen Synonyms:Rpn1|P97|S226S proteasome non-ATPase regulatory subunit 2|non-ATPase|Protein 55.11|proteasome (prosome|26S proteasome regulatory subunit S2|MGC14274,55.11 protein|macropain) 26S subunit|TNFR-associated protein 2|Tumor necrosis factor type 1 receptor-associated protein 2|26S proteasome regulatory subunit RPN1|2|TRAP226S proteasome subunit p97|tumor necrosis factor receptor-associated protein 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PSMD2 (AAH02368, 1 a.a. - 908 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEEGGRDKAPVQPQQSPAAAPGGTDEKPSGKERRDAGDKDKEQELSEEDKQLQDELEMLVERLGEKDTSLYRPALEELRRQIRSSTTSMTSVPKPLKFLRPHYGKLKEIYENMAPGENKRFAADIISVLAMTMSGERECLKYRLVGSQEELASWGHEYVRHLAGEVAKEWQELDDAEKVQREPLLTLVKEIVPYNMAHNAEHEACDLLMEIEQVDMLEKDIDENAYAKVCLYLTSCVNYVPEPENSALLRCALGVFRKFSRFPEALRLALMLNDMELVEDIFTSCKDVVVQKQMAFMLGRHGVFLELSEDVEEYEDLTEIMSNVQLNSNFLALARELDIMEPKVPDDIYKTHLENNRFGGSGSQVDSARMNLASSFVNGFVNAAFGQDKLLTDDGNKWLYKNKDHGMLSAAASLGMILLWDVDGGLTQIDKYLYSSEDYIKSGALLACGIVNSGVRNECDPALALLSDYVLHNSNTMRLGSIFGLGLAYAGSNREDVLTLLLPVMGDSKSSMEVAGVTALACGMIAVGSCNGDVTSTILQTIMEKSETELKDTYARWLPLGLGLNHLGKGEAIEAILAALEVVSEPFRSFANTLVDVCAYAGSGNVLKVQQLLHICSEHFDSKEKEEDKDKKEKKDKDKKEAPADMGAHQGVAVLGIALIAMGEEIGAEMALRTFGHLLRYGEPTLRRAVPLALALISVSNPRLNILDTLSKFSHDADPEVSYNSIFAMGMVGSGTNNARLAAMLRQLAQYHAKDPNNLFMVRLAQGLTHLGKGTLTLCPYHSDRQLMSQVAVAGLLTVLVSFLDVRNIILGKSHYVLYGLVAAMQPRMLVTFDEELRPLPVSVRVGQAVDVVGQAGKPKTITGFQTHTTPVLLAHGERAELATEEFLPVTPILEGFVILRKNPNYDL
- Purification:IgG purified
- Cat. No.:103308-512
- Supplier no.:H00005708-M10
Specifications
About this item
The Proteasome 19S 10B Antibody (1B4) from Novus Biologicals is a mouse monoclonal antibody to Proteasome 19S 10B. This antibody reacts with human. The Proteasome 19S 10B Antibody (1B4) has been validated for the following applications: ELISA.
Type: Primary
Antigen: Proteasome 19S 10B
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human