To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Polycystin 2
- Clonality:Monoclonal
- Clone:4C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:5311
- Antigen Synonyms:Polycystic kidney disease 2 protein|MGC138468|polycystic kidney disease 2 (autosomal dominant)|subfamily P|polycystin-2|Autosomal dominant polycystic kidney disease type II protein|Polycystwin|Pc-2|MGC138466|PC2|PKD4|TRPP2|R48321|member 2|transient receptor potential cation channel|APKD2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PKD2 (NP_000288 261 a.a. - 360 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PVSKTEKTNFKTLSSMEDFWKFTEGSLLDGLYWKMQPSNQTEADNRSFIFYENLLLGVPRIRQLRVRNGSCSIPQDLRDEIKECYDVYSVSSEDRAPFGP
- Purification:IgG purified
- Cat. No.:103307-518
- Supplier no.:H00005311-M01
Specifications
About this item
The Polycystin 2 Antibody (4C8) from Novus Biologicals is a mouse monoclonal antibody to Polycystin 2. This antibody reacts with human. The Polycystin 2 Antibody (4C8) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Polycystin 2
Clonality: Monoclonal
Clone: 4C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human