Anti-UHRF1 Mouse Monoclonal Antibody [clone: 3B12]
103343-042
- Antibody Type:Primary
- Antigen Symbol:UHRF1
- Clonality:Monoclonal
- Clone:3B12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:29128
- Antigen Synonyms:EC 6.3.2.-|FLJ21925|ICBP90NP95|ubiquitin-like with PHD and ring finger domains 1|Ubiquitin-like PHD and RING finger domain-containing protein 1|E3 ubiquitin-protein ligase UHRF1|RNF106MGC138707|ubiquitin-like|Nuclear protein 95|Ubiquitin-like-containing PHD and RING finger domains protein 1|Np95|Nuclear zinc finger protein Np95|huNp95|hNP95|1|Inverted CCAAT box-binding protein of 90 kDa|RING finger protein 106|EC 6.3.2|containing PHD and RING finger domains|Transcription factor ICBP90
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:UHRF1 (NP_037414, 694 a.a. ~ 794 a.a) partial recombinant protein with GST tag.WNEVLASLKDRPASGSPFQLFLSKVEETFQCICCQELVFRPITTVCQHNVCKDCLDRSFRAQVFSCPACRYDLGRSYAMQVNQPLQTVLNQLFPGYGNGR
- Purification:IgG purified
- Cat. No.:103343-042
- Supplier no.:H00029128-M02
The UHRF1 Antibody (3B12) from Novus Biologicals is a mouse monoclonal antibody to UHRF1. This antibody reacts with human. The UHRF1 Antibody (3B12) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: UHRF1
Clonality: Monoclonal
Clone: 3B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human