To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TROY/TNFRSF19
- Clonality:Monoclonal
- Clone:933214
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:25ug
- Gene ID:55504
- Antigen Synonyms:TAJ-alpha|TROYtumor necrosis factor receptor superfamily member 19|TRADE|member 19|tumor necrosis factor receptor superfamily|TAJToxicity and JNK inducer
- Storage Buffer:PBS
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Partial recombinant human TROY/TNFRSF protein (19 aa30-170)
- Tested Applications:, CyTOF-ready
- Purification:Protein A or G purified
- Cat. No.:103295-548
- Supplier no.:MAB1548-SP
Specifications
About this item
The TROY / TNFRSF19 Antibody (933214) - Azide Free from Novus Biologicals is a mouse monoclonal antibody to TROY / TNFRSF19. This antibody reacts with human. The TROY / TNFRSF19 Antibody (933214) - Azide Free has been validated for the following applications: Flow Cytometry, CyTOF-ready.
Type: Primary
Antigen: TROY/TNFRSF19
Clonality: Monoclonal
Clone: 933214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human