Anti-TRF-2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4A794.15]
Catalog # 103359-364
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TRF-2
- Clonality:Monoclonal
- Clone:4A794.15
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Size:100 μl
- Gene ID:7014
- Antigen Synonyms:Telomeric repeat binding protein 2|TRBF 2|TRBF2|telomeric repeat binding factor 2|TERF2|TTAGGG repeat-binding factor 2|TTAGGG repeat binding factor 2|telomeric repeat-binding factor 2|Telomeric DNA-binding protein|TRF 2|TRF2|TERF 2
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Baculovirus expressed whole length TRF2 protein was used for immunizing mice and splenocytes used to generate the hybridoma clone (NP_005643).
- Purification:Protein G purified
- Cat. No.:103359-364
Specifications
About this item
The TRF-2 Antibody (4A794.15) [PE] from Novus Biologicals is a mouse monoclonal antibody to TRF-2. This antibody reacts with human, mouse, rat. The TRF-2 Antibody (4A794.15) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: TRF-2
Clonality: Monoclonal
Clone: 4A794.15
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat