Anti-TRAPA Mouse Polyclonal Antibody
103310-794
- Antibody Type:Primary
- Antigen Name:TRAP alpha
- Antigen Symbol:TRAPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:6745
- Antigen Synonyms:FLJ22100|translocon-associated protein subunit alpha|alpha|TRAPASignal sequence receptor subunit alpha|FLJ78242|SSR alpha subunit|SSR-alpha|FLJ93042|signal sequence receptor|FLJ23034|translocon-associated protein alpha subunit|DKFZp781N23103|TRAP-alpha|FLJ14232|TRAP alpha
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SSR1 (AAH07710.1, 1 a.a. - 286 a.a.) full-length human protein. MRLLPRLLLLLLLVFPATVLFRGGPRGLLAVAQDLTEDEETVEDSIIEDEDDEAEVEEDEPTDLVEDKEEEDVSGEPEASPSADTTILFVKGEDFPANNIVKFLVGFTNKGTEDFIVESLDASFRYPQDYQFYIQNFTALPLNTVVPPQRQATFEYSFIPAEPMGGRPFGLVINLNYKDLNGNVFQDAVFNQTVTVIEREDGLDGETIFMYMFLAGLGLLVIVGLHQLLESRKRKRPIQKVEMGTSSQNDVDMSWIPQETLNQINKASPRRLPRKRAQKRSVGSDE
- Purification:Protein A purified
- Cat. No.:103310-794
- Supplier no.:H00006745-B01P
The TRAP alpha Antibody from Novus Biologicals is a mouse polyclonal antibody to TRAP alpha. This antibody reacts with human. The TRAP alpha Antibody has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: TRAPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human