Anti-TRAF-3 Rabbit Polyclonal Antibody
Catalog # 103360-918
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TRAF-3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.05 ml
- Gene ID:7187
- Antigen Synonyms:CD40bp|CD40-binding protein|CAP-1CD40 receptor associated factor 1|CRAF1CAP1|TNF receptor-associated factor 3|CD40 binding protein|LAP1CD40BP|CD40 receptor-associated factor 1|CD40 associated protein 1|LMP1-associated protein 1
- Storage Buffer:PBS 0.5%BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:A peptide corresponding to amino acids between 300 and 350 of human TRAF3 was used as immunogen for this antibody.
- Purification:Protein G purified
- Cat. No.:103360-918
Specifications
About this item
The TRAF-3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRAF-3. This antibody reacts with human, mouse. The TRAF-3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TRAF-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse