Anti-TPGS2 Mouse Polyclonal Antibody
Catalog # 103341-966
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TPGS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:25941
- Antigen Synonyms:HsT3006|tubulin polyglutamylase complex subunit 2|DKFZp586M1523|L17|PGs2|chromosome 18 open reading frame 10
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:C18orf10 (NP_056291.2, 1 a.a. ~ 300 a.a) full-length human protein.MEEEASSPGLGCSKPHLEKLTLGITRILESSPGVTEVTIIEKPPAERHMISSWEQKNNCVMPEDVKNFYLMTNGFHMTWSVKLDEHIIPLGSMAINSISKLTQLTQSSMYSLPNAPTLADLEDDTHEASDDQPEKPHFDSRSVIFELDSCNGSGKVCLVYKSGKPALAEDTEIWFLDRALYWHFLTDTFTAYYRLLITHLGLPQWQYAFTSYGISPQAKQWFSMYKPITYNTNLLTEETDSFVNKLDPSKVFKSK
- Purification:Protein A purified
- Cat. No.:103341-966
- Supplier no.:H00025941-B01P
Specifications
About this item
The TPGS2 Antibody from Novus Biologicals is a mouse polyclonal antibody to TPGS2. This antibody reacts with human. The TPGS2 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: TPGS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human