Anti-TNFRSF11B Mouse Monoclonal Antibody [clone: 98A1071]
Catalog # 103359-340
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Osteoprotegerin
- Antigen Symbol:TNFRSF11B
- Clonality:Monoclonal
- Clone:98A1071
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:0.025 mg
- Gene ID:4982
- Antigen Synonyms:Osteoprotegerin|TR1|tumor necrosis factor receptor superfamily|member 11b|Osteoclastogenesis inhibitory factor|OCIFMGC29565|OPGtumor necrosis factor receptor superfamily member 11B
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:A peptide corresponding to the amino acids 20-37 TQETFPPKYLHYDEETSH of human Osteoprotegerin was used as immunogen for this antibody.
- Tested Applications:CyTOF-ready
- Purification:Protein G purified
- Cat. No.:103359-340
Specifications
About this item
The Osteoprotegerin / TNFRSF11B Antibody (98A1071) from Novus Biologicals is a mouse monoclonal antibody to Osteoprotegerin / TNFRSF11B. This antibody reacts with human, rat. The Osteoprotegerin / TNFRSF11B Antibody (98A1071) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Type: Primary
Antigen: TNFRSF11B
Clonality: Monoclonal
Clone: 98A1071
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat