To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TLR6
- Clonality:Monoclonal
- Clone:86B1153.2
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:10333
- Antigen Synonyms:CD286 antigen|toll-like receptor 6|CD286
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:This antibody was developed against a synthetic peptide corresponding to amino acids 408-424 of human TRL6.
- Purification:Protein G purified
- Cat. No.:103359-916
Specifications
About this item
The TLR6 Antibody (86B1153.2) [PerCP] from Novus Biologicals is a mouse monoclonal antibody to TLR6. This antibody reacts with human. The TLR6 Antibody (86B1153.2) [PerCP] has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TLR6
Clonality: Monoclonal
Clone: 86B1153.2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human