To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TLK2
- Clonality:Monoclonal
- Clone:2G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:11011
- Antigen Synonyms:PKU-ALPHA|MGC44450|EC 2.7.11.1|tousled-like kinase 2serine/threonine kinase|EC 2.7.11|serine/threonine-protein kinase tousled-like 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TLK2 (AAH44925, 1 a.a. ~ 126 a.a) partial recombinant protein with GST tag.MEELHSLDPRRQELLEARFTGVGVSKGPLNSESSNQSLCSVG SLSDKEVETPEKKQNDQRNRKRKAEPYETSQGKGTPRGHKIS DYFEFAGGSAPGTSPGRSVPPVARSSPQHSLSNPLPRRVEQP
- Purification:IgG purified
- Cat. No.:103339-950
- Supplier no.:H00011011-M01
Specifications
About this item
The TLK2 Antibody (2G7) from Novus Biologicals is a mouse monoclonal antibody to TLK2. This antibody reacts with human. The TLK2 Antibody (2G7) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TLK2
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human