Anti-TFAP4 Mouse Monoclonal Antibody [clone: 6B1]
103332-982
- Antibody Type:Primary
- Antigen Symbol:TFAP4
- Clonality:Monoclonal
- Clone:6B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:7023
- Antigen Synonyms:transcription factor AP-4 (activating enhancer binding protein 4)|AP-4|transcription factor AP-4 (activating enhancer-binding protein 4)|Activating enhancer-binding protein 4|bHLHc41Class C basic helix-loop-helix protein 41|BHLHC41|transcription factor AP-4
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TFAP4 (NP_003214, 93 a.a. - 192 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEKAEDLRREMIELRQQLDKERSVRMMLEEQVRSLEAHMYPEKLKVIA
- Purification:IgG purified
- Cat. No.:103332-982
- Supplier no.:H00007023-M01
The TFAP4 Antibody (6B1) from Novus Biologicals is a mouse monoclonal antibody to TFAP4. This antibody reacts with human. The TFAP4 Antibody (6B1) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TFAP4
Clonality: Monoclonal
Clone: 6B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human