To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TEM7/PLXDC1
- Clonality:Monoclonal
- Clone:197C193 (IM193)
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:57125
- Antigen Synonyms:Tumor endothelial marker 7|TEM7DKFZp686F0937|plexin domain containing 1,2410003I07Rik|TEM3FLJ36270|FLJ45632|Tumor endothelial marker 3|plexin domain-containing protein 1
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Amino acids 409-425 (LQNNLSPKTKGTPVHLG) of human TEM7 were used to develop this monoclonal antibody.
- Purification:Protein G purified
- Cat. No.:103360-240
Specifications
About this item
The TEM7 / PLXDC1 Antibody (197C193 (IM193)) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to TEM7 / PLXDC1. This antibody reacts with human, mouse, rat. The TEM7 / PLXDC1 Antibody (197C193 (IM193)) [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: TEM7/PLXDC1
Clonality: Monoclonal
Clone: 197C193 (IM193)
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat