Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-STK38 Mouse Monoclonal Antibody [clone: 2F6]
Anti-STK38 Mouse Monoclonal Antibody [clone: 2F6]
Catalog # 103340-580
Supplier:  Novus Biologicals
Anti-STK38 Mouse Monoclonal Antibody [clone: 2F6]
Catalog # 103340-580
Supplier:  Novus Biologicals
Supplier Number:  H00011329-M11

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    STK38
  • Clonality:
    Monoclonal
  • Clone:
    2F6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    11329
  • Antigen Synonyms:
    NDRnuclear Dbf2-related 1|NDR1 protein kinase|NDR1|serine threonine protein kinase|serine/threonine-protein kinase 38|EC 2.7.11.1|EC 2.7.11|Nuclear Dbf2-related kinase 1|Ndr Ser/Thr kinase-like protein|serine/threonine kinase 38
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    STK38 (AAH12085, 365 a.a. ~ 466 a.a) partial recombinant protein with GST tag. EHRIGAPGVEEIKSNSFFEGVDWEHIRERPAAISIEIKSIDDTSNFDEFPESDILKPTVATSNHPETDYKNKDWVFINYTYKRFEGLTARGAIPSYMKAAK
  • Purification:
    IgG purified
  • Cat. No.:
    103340-580
  • Supplier no.:
    H00011329-M11

Specifications

About this item

The STK38 Antibody (2F6) from Novus Biologicals is a mouse monoclonal antibody to STK38. This antibody reacts with human. The STK38 Antibody (2F6) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STK38
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human