Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-STK38 Mouse Monoclonal Antibody [clone: 3A5]
Anti-STK38 Mouse Monoclonal Antibody [clone: 3A5]
Catalog # 103340-572
Supplier:  Novus Biologicals
Anti-STK38 Mouse Monoclonal Antibody [clone: 3A5]
Catalog # 103340-572
Supplier:  Novus Biologicals
Supplier Number:  H00011329-M02

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    STK38
  • Clonality:
    Monoclonal
  • Clone:
    3A5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    11329
  • Antigen Synonyms:
    NDRnuclear Dbf2-related 1|NDR1 protein kinase|NDR1|serine threonine protein kinase|serine/threonine-protein kinase 38|EC 2.7.11.1|EC 2.7.11|Nuclear Dbf2-related kinase 1|Ndr Ser/Thr kinase-like protein|serine/threonine kinase 38
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    STK38 (AAH12085, 365 a.a. ~ 466 a.a) partial recombinant protein with GST tag.EHRIGAPGVEEIKSNSFFEGVDWEHIRERPAAISIEIKSIDDTSNFDEFPESDILKPTVATSNHPETDYKNKDWVFINYTYKRFEGLTARGAIPSYMKAAK
  • Purification:
    IgG purified
  • Cat. No.:
    103340-572
  • Supplier no.:
    H00011329-M02

Specifications

About this item

The STK38 Antibody (3A5) from Novus Biologicals is a mouse monoclonal antibody to STK38. This antibody reacts with human, mouse, rat. The STK38 Antibody (3A5) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STK38
Clonality: Monoclonal
Clone: 3A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat