Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SQLE Rabbit Polyclonal Antibody
Anti-SQLE Rabbit Polyclonal Antibody
Catalog # 103287-272
Supplier:  Novus Biologicals
Anti-SQLE Rabbit Polyclonal Antibody
Catalog # 103287-272
Supplier:  Novus Biologicals
Supplier Number:  NBP2-32001

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    SQLE
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Size:
    100 μl
  • Gene ID:
    6713
  • Antigen Synonyms:
    squalene epoxidaseSE|FLJ30795|squalene monooxygenase|ERG1|EC 1.14.99.7
  • Storage Buffer:
    PBS (pH 7.2) and 40% Glycerol
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against a recombinant protein corresponding to amino acids: DGRKVTVIERDLKEPDRIVGEFLQPGGYHVLKDLGLGDTVEGLDAQVVNGYMIHDQESKSEVQIPYPLSENNQVQSGRAFHHGRFIMSLRKAAMAEPNAKFIEGVVLQLLEEDDVVMG
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103287-272
  • Supplier no.:
    NBP2-32001

Specifications

About this item

The SQLE Antibody from Novus Biologicals is a rabbit polyclonal antibody to SQLE. This antibody reacts with human, mouse. The SQLE Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SQLE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse