To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:SLC31A1/CTR1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 405
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Pig,Xenopus,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1317
- Antigen Synonyms:solute carrier family 31 (copper transporters)|Solute carrier family 31 member 1|CTR1MGC75487|Copper transporter 1|hCTR1copper transport 1 homolog|COPT1high affinity copper uptake protein 1|member 1
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide derived from a C-terminal sequence of human SLC31A1/CTR1 [UniProt# O15431]
- Purification:Immunogen affinity purified
- Cat. No.:103357-970
The SLC31A1 / CTR1 Antibody [Alexa Fluor« 405] from Novus Biologicals is a rabbit polyclonal antibody to SLC31A1 / CTR1. This antibody reacts with human, mouse, rat, porcine, xenopus, zebrafish. The SLC31A1 / CTR1 Antibody [Alexa Fluor« 405] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: SLC31A1/CTR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 405
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Xenopus, Zebrafish