Anti-RSAD1 Rabbit Polyclonal Antibody
Catalog # 103258-734
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:RSAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:55316
- Antigen Synonyms:radical S-adenosyl methionine domain-containing protein 1|FLJ11164|FLJ20975|Oxygen-independent coproporphyrinogen-III oxidase-like protein RSAD1|mitochondrial|radical S-adenosyl methionine domain containing 1|EC 1.3.99.-|EC 1.3.99|EC 1.3.99.22
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:VTLEANPTSAPGSRLAEFGAAGVNRLSIGLQSLDDTELRLLGRTHSACDALRTLAEARRLFPGRVSVDLMLGLPAQQVGPWLGQLQELLHHC
- Purification:Immunogen affinity purified
- Cat. No.:103258-734
- Supplier no.:NBP1-83771
Specifications
About this item
The RSAD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RSAD1. This antibody reacts with human. The RSAD1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: RSAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human