Anti-RPE65 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 401.8B11.3D9]
Catalog # 103357-864
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:RPE65
- Clonality:Monoclonal
- Clone:401.8B11.3D9
- Conjugation:Alexa Fluor® 647
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Pig,Xenopus,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:0.2 ml
- Gene ID:6121
- Antigen Synonyms:sRPE65|LCA2p63|All-trans-retinyl-palmitate hydrolase|EC 3.1.1.64|EC 5.2.1.7|Retinal pigment epithelium-specific 65 kDa protein|retinoid isomerohydrolase|retinitis pigmentosa 20 (autosomal recessive)|RBP-binding membrane protein|retinal pigment epithelium-specific protein 65kDa|Retinol isomerase|retinal pigment epithelium-specific protein (65kD)|RP20|rd12|mRPE65
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Bovine RPE65 microsomal membrane proteins. [UniProt# Q28175]
- Purification:Protein G purified
- Cat. No.:103357-864
Specifications
About this item
The RPE65 Antibody (401.8B11.3D9) [Alexa Fluor« 647] from Novus Biologicals is a mouse monoclonal antibody to RPE65. This antibody reacts with human, mouse, rat, porcine, bovine, canine, chicken, xenopus. The RPE65 Antibody (401.8B11.3D9) [Alexa Fluor« 647] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: RPE65
Clonality: Monoclonal
Clone: 401.8B11.3D9
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Xenopus