Anti-RPC62 Mouse Polyclonal Antibody
Catalog # 103339-294
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:RPC62
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:10623
- Antigen Synonyms:DNA-directed RNA polymerase III subunit RPC3|DNA-directed RNA polymerase III subunit C|DNA-directed III 62 kDa polypeptide|RPC62RPC3|RNA polymerase III 62 kDa subunit|polymerase (RNA) III (DNA directed) polypeptide C (62kD)|RNA polymerase III subunit C3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:POLR3C (NP_006459.3, 1 a.a. ~ 534 a.a) full-length human protein.MTQAEIKLCSLLLQEHFGEIVEKIGVHLIRTGSQPLRVIAHDTGTSLDQVKKALCVLVQHNLVSYQVHKRGVVEYEAQCSRVLRMLRYPRYIYTTKTLYSDTGELIVEELLLNGKLTMSAVVKKVADRLTETMEDGKTMDYAEVSNTFVRLADTHFVQRCPSVPTTENSDPGPPPPAPTLVINEKDMYLVPKLSLIGKGKRRRSSDEDAAGEPKAKRPKYTTDNKEPIPDDGIYWQANLDRFHQHFRDQAIVSAV
- Purification:Protein A purified
- Cat. No.:103339-294
- Supplier no.:H00010623-B03P
Specifications
About this item
The RPC62 Antibody from Novus Biologicals is a mouse polyclonal antibody to RPC62. This antibody reacts with human, rat. The RPC62 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: RPC62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat