To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:RAB11FIP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:26056
- Antigen Synonyms:GAF1rab11-FIP5|Phosphoprotein pp75|pp75|RAB11RIP5|Rab11-FIP5|Gamma-SNAP-associated factor 1|DKFZp434H018|RIP11rab11 family-interacting protein 5|Rab11-interacting protein Rip11|KIAA0857gaf-1|Gaf-1|RAB11 family interacting protein 5 (class I)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:RAB11FIP5 (AAH35013, 1 a.a. ~ 652 a.a) full-length human protein.MALVRGAEPAAGPSRWLPTHVQVTVLRARGLRGKSSGAGSTSDAYTVIQVGREKYSTSVVEKTHGCPEWREECSFELPPGALDGLLRAQEADAGPAPWAASSAAACELVLTTMHRSLIGVDKFLGQATVALDEVFGAGRAQHTQWYKLHSKPGKKEKERGEIEVTIQFTRNNLSAMFDLSMKDKPRSPFSKIRDKMKGKKKYDLESASAILPSSAIEDPDLGSLGKMGKAKGFFLRNKLRKSSLTQSNTSLGSDS
- Purification:Protein A purified
- Cat. No.:103342-050
- Supplier no.:H00026056-B01P
Specifications
About this item
The RAB11FIP5 Antibody from Novus Biologicals is a mouse polyclonal antibody to RAB11FIP5. This antibody reacts with human. The RAB11FIP5 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: RAB11FIP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human